2-Amino-5-pyrimidineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminopyrimidine-5-boronic acid, (2-Aminopyrimidin-5-yl)boronic acid

A172972-1G A172972-250MG A172972-25G A172972-5G
77273-772EA 6.18 USD
77273-772 77273-774 77273-776 77273-778
2-Amino-5-pyrimidineboronic acid 98%
2-Amino-5-pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₄H₆BN₃O₂
MW: 138.92 g/mol
MDL Number: MFCD07375147
CAS Number: 936250-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR