2-Ethyl-4-hydroxy-5-methyl-3(2H)-furanone 95%

Supplier: Ambeed
Warning

Synonyms: 2-Ethyl-4-hydroxy-5-methylfuran-3(2H)-one, Ethyl furaneol

A172960-100G A172960-1G A172960-25G A172960-5G
77259-988EA 88.58 USD
77259-988 77259-990 77259-992 77259-994
2-Ethyl-4-hydroxy-5-methyl-3(2H)-furanone 95%
2-Ethyl-4-hydroxy-5-methyl-3(2H)-furanone

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MW: 142.15 g/mol
MDL Number: MFCD00191360
CAS Number: 27538-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR