4'-((4-Chlorophenyl)sulfonyl)-[1,1'-biphenyl]-4-ol 97%

Supplier: Ambeed
Warning

A172946-1G A172946-250MG A172946-25G A172946-5G
77248-076EA 63 USD
77248-076 77248-078 77248-080 77248-082
4'-((4-Chlorophenyl)sulfonyl)-[1,1'-biphenyl]-4-ol 97%
4'-((4-Chlorophenyl)sulfonyl)-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₈H₁₃ClO₃S
MDL Number: MFCD30543160
CAS Number: 30809-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR