CRAC Channel Inhibitor, BTP2 98%

Supplier: Ambeed
Warning

A172938-100MG A172938-10MG A172938-1MG A172938-25MG A172938-2MG A172938-50MG A172938-5MG
77259-974EA 370 USD
77259-974 77259-976 77259-978 77259-980 77259-982 77259-984 77259-986
CRAC Channel Inhibitor, BTP2 98%
CRAC Channel Inhibitor, BTP2

Precaución: for Research Use Only

Formula: C₁₅H₉F₆N₅OS
MDL Number: MFCD00220976
CAS Number: 223499-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR