Di-2-thienyl ketone 97%

Supplier: Ambeed
Warning

A172873-100MG A172873-10G A172873-1G A172873-250MG A172873-5G
77259-964EA 5 USD
77259-964 77259-966 77259-968 77259-970 77259-972
Di-2-thienyl ketone 97%
Di-2-thienyl ketone

Precaución: for Research Use Only

Formula: C₉H₆OS₂
MDL Number: MFCD00052096
CAS Number: 704-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR