N-Benzyl-N-butyl-3-methylaniline 97%

Supplier: Ambeed
Warning

A172856-100G A172856-10G A172856-25G
77236-312EA 26.78 USD
77236-312 77236-314 77236-316
N-Benzyl-N-butyl-3-methylaniline 97%
N-Benzyl-N-butyl-3-methylaniline

Precaución: for Research Use Only

Formula: C₁₈H₂₃N
MW: 253.38 g/mol
MDL Number: MFCD04038164
CAS Number: 187101-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR