3-Bromo-2-iodophenol 97%

Supplier: Ambeed
Warning

A172688-100MG A172688-10G A172688-1G A172688-250MG A172688-25G
77273-762EA 14 USD
77273-762 77273-764 77273-766 77273-768 77273-770
3-Bromo-2-iodophenol 97%
3-Bromo-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
MDL Number: MFCD08166320
CAS Number: 855836-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR