3-Aminopiperidine-2,6-dione hydrobromide 97%

Supplier: Ambeed
Warning

A172642-10G A172642-1G A172642-5G
77248-066EA 63 USD
77248-066 77248-068 77248-070
3-Aminopiperidine-2,6-dione hydrobromide 97%
3-Aminopiperidine-2,6-dione hydrobromide

Precaución: for Research Use Only

Formula: C₅H₉BrN₂O₂
MW: 209.04 g/mol
MDL Number: MFCD30573975
CAS Number: 90802-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR