Boc-D-Leu-OMe 97%

Supplier: Ambeed
Warning

A172586-100G A172586-10G A172586-1G A172586-25G A172586-5G
77248-052EA 226.6 USD
77248-052 77248-054 77248-056 77248-058 77248-060
Boc-D-Leu-OMe 97%
Boc-D-Leu-OMe

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₄
MW: 245.31 g/mol
MDL Number: MFCD11041224
CAS Number: 133467-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR