(2,3-Dichlorophenoxy)acetic acid 97%

Supplier: Ambeed
Warning

A172507-25G A172507-5G
77248-050EA 67 USD
77248-050 77746-140
(2,3-Dichlorophenoxy)acetic acid 97%
(2,3-Dichlorophenoxy)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃Cl₂)OCH₂COOH
MDL Number: MFCD00004299
CAS Number: 2976-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR