4-((3-Methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Isoimperatorin
A172258-10MG
A172258-1MG
A172258-25MG
A172258-5MG
77273-752EA
12.36
USD
77273-752
77273-754
77273-756
77273-758
4-((3-Methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one 95%
4-((3-Methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one
Precaución: for Research Use Only
Formula:
C₁₆H₁₄O₄ MW: 270.28 g/mol |
MDL Number:
MFCD00272155 CAS Number: 482-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...