6-(4-Methylphenyl)nicotinaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 6-(p-Tolyl)nicotinaldehyde

A172161-100MG A172161-250MG
77236-296EA 74.16 USD
77236-296 77236-298
6-(4-Methylphenyl)nicotinaldehyde 97%
6-(4-Methylphenyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
MDL Number: MFCD04115442
CAS Number: 898795-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR