2,6-Dichloro-N1-(4,5-dihydro-1H-imidazol-2-yl)benzene-1,4-diamine hydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: Apraclonidine HCl

A172137-1MG A172137-25MG
77248-034EA 5.15 USD
77248-034 77248-036
2,6-Dichloro-N1-(4,5-dihydro-1H-imidazol-2-yl)benzene-1,4-diamine hydrochloride 97%
2,6-Dichloro-N1-(4,5-dihydro-1H-imidazol-2-yl)benzene-1,4-diamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁Cl₃N₄
MW: 281.57 g/mol
MDL Number: MFCD00135922
CAS Number: 73218-79-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR