Trimethylpropylammonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: N,N,N-Trimethylpropan-1-aminium bromide

A172028-1G A172028-5G A172028-100G
77248-030EA 8 USD
77248-030 77248-032 77567-530
Trimethylpropylammonium bromide 97%
Trimethylpropylammonium bromide

Precaución: For research use only.

Formula: C₆H₁₆BrN
MDL Number: MFCD06227832
CAS Number: 2650-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR