Methyl-2-cyanoisonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-cyanoisonicotinate

A171956-100G A171956-10G A171956-1G A171956-25G A171956-5G
77259-944EA 226.6 USD
77259-944 77259-946 77259-948 77259-950 77259-952
Methyl-2-cyanoisonicotinate 98%
Methyl-2-cyanoisonicotinate

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.148 g/mol
MDL Number: MFCD07367894
CAS Number: 94413-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR