4-Amino-3-hydroxybenzonitrile 95%
Supplier: Ambeed
|
Danger
|
A171916-100MG
A171916-250MG
A171916-1G
77286-908EA
20
USD
77286-908
77286-910
77567-528
4-Amino-3-hydroxybenzonitrile 95%
4-Amino-3-hydroxybenzonitrile
Precaución: For research use only.
Formula:
C₇H₆N₂O |
MDL Number:
MFCD06657033 CAS Number: 55586-26-0 UN: 3439 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...