3-(4-Bromophenyl)propiononitrile 95%

Supplier: Ambeed
Warning

A171809-100MG A171809-10G A171809-1G A171809-250MG A171809-5G
77259-934EA 14 USD
77259-934 77259-936 77259-938 77259-940 77259-942
3-(4-Bromophenyl)propiononitrile 95%
3-(4-Bromophenyl)propiononitrile

Precaución: for Research Use Only

Formula: C₉H₈BrN
MDL Number: MFCD01310795
CAS Number: 57775-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR