2,3-Norbornanedicarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Bicyclo[2.2.1]heptane-2,3-dicarboxylic Acid

A171748-1G A171748-5G
77248-026EA 19.57 USD
77248-026 77248-028
2,3-Norbornanedicarboxylic acid 98%
2,3-Norbornanedicarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₂O₄
MW: 184.19 g/mol
MDL Number: MFCD00520577
CAS Number: 1724-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR