2-Methylquinoline-6-carboxylic acid 98%

Supplier: Ambeed
Warning

A171706-10G A171706-5G A171706-25G
77273-728EA 15 USD
77273-728 77273-730 77748-138
2-Methylquinoline-6-carboxylic acid 98%
2-Methylquinoline-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MDL Number: MFCD00748504
CAS Number: 635-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR