3-Bromo-5-nitropyridin-4-amine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-bromo-5-nitropyridine

A171612-100MG A171612-10G A171612-250MG A171612-5G A171612-1G A171612-25G
77286-880EA 5.15 USD
77286-880 77286-882 77286-884 77748-137 77748-135 77748-136
3-Bromo-5-nitropyridin-4-amine 98%
3-Bromo-5-nitropyridin-4-amine

Precaución: for Research Use Only

Formula: C5H4BrN3O2
MW: 218.0100 g/mol
MDL Number: MFCD01646057
CAS Number: 89284-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR