3'-Chloro-4'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

A171552-1G A171552-250MG
77236-282EA 12 USD
77236-282 77236-284
3'-Chloro-4'-hydroxyacetophenone 98%
3'-Chloro-4'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MDL Number: MFCD00663621
CAS Number: 2892-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR