Icariin 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

A171476-100MG A171476-10G A171476-1G A171476-250MG A171476-25MG A171476-5G
77286-868EA 7.21 USD
77286-868 77286-870 77286-872 77286-874 77286-876 77286-878
Icariin 98%
Icariin

Precaución: for Research Use Only

Formula: C₃₃H₄₀O₁₅
MW: 676.67 g/mol
MDL Number: MFCD00210516
CAS Number: 489-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR