4-(2-(2-Cyclopentylidenehydrazinyl)thiazol-4-yl)benzonitrile hydrobromide 98%

Supplier: Ambeed
Warning

A171428-10MG A171428-1MG A171428-25MG A171428-50MG A171428-5MG
77236-272EA 67 USD
77236-272 77236-274 77236-276 77236-278 77236-280
4-(2-(2-Cyclopentylidenehydrazinyl)thiazol-4-yl)benzonitrile hydrobromide 98%
4-(2-(2-Cyclopentylidenehydrazinyl)thiazol-4-yl)benzonitrile hydrobromide

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrN₄S
MDL Number: MFCD28137688
CAS Number: 1622921-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR