D-Phenylalaninol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
D-Penylalaninol, D(+)-2-Amino-3-phenyl-1-propanol
A171066-100G
A171066-10G
A171066-1G
A171066-25G
A171066-500G
A171066-5G
77247-998EA
113.3
USD
77247-998
77248-000
77248-002
77248-004
77248-006
77248-008
D-Phenylalaninol 98%
D-Phenylalaninol
Precaución: for Research Use Only
Formula:
C₉H₁₃NO MW: 151.2 g/mol |
MDL Number:
MFCD00064298 CAS Number: 5267-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...