2,4,6-Tribromobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A171056-1G
A171056-250MG
A171056-25G
A171056-5G
77247-990EA
44
USD
77247-990
77247-992
77247-994
77247-996
2,4,6-Tribromobenzoic acid 95%
2,4,6-Tribromobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃Br₃O₂ |
MDL Number:
MFCD00059882 CAS Number: 633-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...