4'-Nitroacetanilide 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Nitrophenyl)acetamide

A171029-100G A171029-25G A171029-5G
77247-984EA 114.33 USD
77247-984 77247-986 77247-988
4'-Nitroacetanilide 98%
4'-Nitroacetanilide

Precaución: for Research Use Only

Formula: C₈H₈N₂O₃
MW: 180.16 g/mol
MDL Number: MFCD00007303
CAS Number: 104-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR