3-phenylimidazolidine-2,4-dione 95%

Supplier: Ambeed
Warning

Synonyms: 3-Phenylimidazolidine-2,4-dione, 3-Phenylhydantoin

A170995-100MG A170995-1G A170995-250MG A170995-5G
77286-850EA 40.17 USD
77286-850 77286-852 77286-854 77286-856
3-phenylimidazolidine-2,4-dione 95%
3-phenylimidazolidine-2,4-dione

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.175 g/mol
MDL Number: MFCD00209486
CAS Number: 2221-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR