2-(2,2-Dimethyl-1,3-dioxolan-4-yl)ethanol 90%

Supplier: Ambeed
Warning

A170712-10G A170712-1G A170712-25G A170712-5G
77286-828EA 100 USD
77286-828 77286-830 77286-832 77286-834
2-(2,2-Dimethyl-1,3-dioxolan-4-yl)ethanol 90%
2-(2,2-Dimethyl-1,3-dioxolan-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₁₄O₃
MDL Number: MFCD03701622
CAS Number: 5754-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR