4-(4-Chlorobenzyl)benzene-1,3-diol 97%

Supplier: Ambeed
Warning

A170679-100MG A170679-1G A170679-250MG
77247-978EA 53.56 USD
77247-978 77247-980 77247-982
4-(4-Chlorobenzyl)benzene-1,3-diol 97%
4-(4-Chlorobenzyl)benzene-1,3-diol

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO₂
MW: 234.67 g/mol
MDL Number: MFCD28969120
CAS Number: 6280-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR