D-Methionine 98%

Supplier: Ambeed
Warning

Synonyms: D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH

A170637-10G A170637-25G A170637-500G A170637-5G A170637-100G
77286-820EA 8.24 USD
77286-820 77286-822 77286-824 77286-826 77748-129
D-Methionine 98%
D-Methionine

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
MDL Number: MFCD00002622
CAS Number: 348-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR