2,3-Dihydrobenzo[1,4]dioxine-5-carbonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-5-carbonyl chloride

A170453-1G A170453-250MG
77247-974EA 127.72 USD
77247-974 77247-976
2,3-Dihydrobenzo[1,4]dioxine-5-carbonyl chloride 95%
2,3-Dihydrobenzo[1,4]dioxine-5-carbonyl chloride

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
MW: 198.61 g/mol
MDL Number: MFCD04972616
CAS Number: 38871-41-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR