N-Fmoc-L-alanine 98% (max.6% H₂O)

Supplier: Ambeed
Warning

Synonyms: N-(9-Fluorenylmethoxycarbonyl)-L-alanine, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine, Fmoc-L-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine, (-)-L-Alanine, N-FMOC protected, (-)-FMOC-L-Ala-OH, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected, Fmoc-Ala-OH

A170416-1000G A170416-100G A170416-10G A170416-1G A170416-25G A170416-500G A170416-5G
77273-702EA 267 USD
77273-702 77273-704 77273-706 77273-708 77273-710 77273-712 77273-714
N-Fmoc-L-alanine 98% (max.6% H₂O)
N-Fmoc-L-alanine

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₄
MDL Number: MFCD00037139
CAS Number: 35661-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR