Ethyl-4-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluorobenzoic acid ethyl ester, Ethyl 4-fluorobenzoate

A170402-1000G A170402-100G A170402-10G A170402-25G A170402-500G A170402-5G
77273-690EA 140.08 USD
77273-690 77273-692 77273-694 77273-696 77273-698 77273-700
Ethyl-4-fluorobenzoate 98%
Ethyl-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.17 g/mol
MDL Number: MFCD00000351
CAS Number: 451-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR