(5R,5aR,8aR,9R)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 4'-Demethylpodophyllotoxin

A170380-10MG A170380-1MG A170380-25MG A170380-50MG
77286-812EA 80.34 USD
77286-812 77286-814 77286-816 77286-818
(5R,5aR,8aR,9R)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one 97%
(5R,5aR,8aR,9R)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one

Precaución: for Research Use Only

Formula: C₂₁H₂₀O₈
MW: 400.37 g/mol
MDL Number: MFCD01725607
CAS Number: 40505-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR