4-Bromoisoindoline hydrochloride 95%

Supplier: Ambeed
Warning

A170251-100MG A170251-10G A170251-1G A170251-250MG A170251-25G A170251-5G
77286-800EA 7 USD
77286-800 77286-802 77286-804 77286-806 77286-808 77286-810
4-Bromoisoindoline hydrochloride 95%
4-Bromoisoindoline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉BrClN
MDL Number: MFCD10700134
CAS Number: 923590-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR