3-(Aminomethyl)-5-bromopyridine 98%

Supplier: Ambeed
Warning

A170215-100MG A170215-1G A170215-250MG A170215-5G
77273-682EA 8 USD
77273-682 77273-684 77273-686 77273-688
3-(Aminomethyl)-5-bromopyridine 98%
3-(Aminomethyl)-5-bromopyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MDL Number: MFCD06212876
CAS Number: 135124-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR