2-[Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]benzaldehyde 98%

Supplier: Ambeed
Warning

A170130-1G A170130-250MG
77247-958EA 675 USD
77247-958 77247-960
2-[Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]benzaldehyde 98%
2-[Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]benzaldehyde

Precaución: for Research Use Only

Formula: C₃₇H₅₁O₃P
MDL Number: MFCD15143663
CAS Number: 1202865-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR