1-Ethyl-3-(2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea 98%

Supplier: Ambeed
Warning

A170082-100MG A170082-1G A170082-250MG
77286-786EA 36.05 USD
77286-786 77286-788 77286-790
1-Ethyl-3-(2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea 98%
1-Ethyl-3-(2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea

Precaución: for Research Use Only

Formula: C₁₆H₂₅BN₂O₄
MW: 320.19 g/mol
MDL Number: MFCD08458185
CAS Number: 917111-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR