Methyl 2-(1,2,3,4-tetrahydroquinolin-6-yl)acetate 97%

Supplier: Ambeed
Warning

A169951-100MG A169951-1G
77236-232EA 188.49 USD
77236-232 77236-234
Methyl 2-(1,2,3,4-tetrahydroquinolin-6-yl)acetate 97%
Methyl 2-(1,2,3,4-tetrahydroquinolin-6-yl)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MW: 205.25 g/mol
MDL Number: MFCD24558334
CAS Number: 5622-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR