BMS-303141 98%

Supplier: Ambeed
Warning

A169881-10MG A169881-1MG A169881-5MG A169881-50MG
77247-944EA 74.99 USD
77247-944 77247-946 77247-948 77746-132
BMS-303141 98%
BMS-303141

Precaución: for Research Use Only

Formula: C₁₉H₁₅Cl₂NO₄S
MDL Number: MFCD25976797
CAS Number: 943962-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR