2-Bromo-3-hydroxy-4-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A169768-10G A169768-1G A169768-250MG A169768-25G A169768-5G
77236-222EA 105.06 USD
77236-222 77236-224 77236-226 77236-228 77236-230
2-Bromo-3-hydroxy-4-methoxybenzaldehyde 97%
2-Bromo-3-hydroxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD02380230
CAS Number: 2973-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR