4-(Trifluoromethyl)-1H-imidazole 98%

Supplier: Ambeed
Warning

A169690-100G A169690-10G A169690-1G A169690-250MG A169690-5G A169690-25G
77286-768EA 792 USD
77286-768 77286-770 77286-772 77286-774 77286-776 77567-490
4-(Trifluoromethyl)-1H-imidazole 98%
4-(Trifluoromethyl)-1H-imidazole

Precaución: For research use only.

Formula: C₄H₃F₃N₂
MDL Number: MFCD08458860
CAS Number: 33468-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR