(4-Bromo-2-methylphenyl)methanamine 97%

Supplier: Ambeed
Warning

A169649-100MG A169649-10G A169649-1G A169649-250MG A169649-5G
77286-758EA 27.81 USD
77286-758 77286-760 77286-762 77286-764 77286-766
(4-Bromo-2-methylphenyl)methanamine 97%
(4-Bromo-2-methylphenyl)methanamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.07 g/mol
MDL Number: MFCD01310789
CAS Number: 376646-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR