2-(2-(4-(3-(2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene)propyl)piperazin-1-yl)ethoxy)ethanol dihydrochloride 97%

Supplier: Ambeed
Warning

A169550-100MG A169550-250MG
77236-218EA 61 USD
77236-218 77236-220
2-(2-(4-(3-(2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene)propyl)piperazin-1-yl)ethoxy)ethanol dihydrochloride 97%
2-(2-(4-(3-(2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene)propyl)piperazin-1-yl)ethoxy)ethanol dihydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₃₁Cl₂F₃N₂O₂S
MW: 551.49 g/mol
MDL Number: MFCD30478748
CAS Number: 1535-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR