Sodium ((2R,3R,4S,5R)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl)sulfamate 95%

Supplier: Ambeed
Warning

A169434-100MG A169434-25G A169434-100G
77259-902EA 6 USD
77259-902 77615-414 77748-127
Sodium ((2R,3R,4S,5R)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl)sulfamate 95%
Sodium ((2R,3R,4S,5R)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl)sulfamate

Precaución: for Research Use Only

Formula: C₆H₁₂NNaO₈S
MDL Number: MFCD09832124
CAS Number: 38899-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR