4-(Methoxycarbonylamino)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-((Methoxycarbonyl)amino)phenyl)boronic acid

A169411-1G A169411-5G A169411-250MG
77286-748EA 208 USD
77286-748 77286-750 77748-126
4-(Methoxycarbonylamino)benzeneboronic acid 98%
4-(Methoxycarbonylamino)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₄
MW: 194.98 g/mol
MDL Number: MFCD03788425
CAS Number: 850567-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR