2-Chloro-4-methylbenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A169367-100G
A169367-10G
A169367-1G
A169367-250MG
A169367-25G
A169367-5G
77259-890EA
202
USD
77259-890
77259-892
77259-894
77259-896
77259-898
77259-900
2-Chloro-4-methylbenzonitrile 97%
2-Chloro-4-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆CIN |
MDL Number:
MFCD03094672 CAS Number: 21423-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...