Avibactam Sodium 98%

Supplier: Ambeed
Warning

A169351-100MG A169351-10MG A169351-1G A169351-1MG A169351-50MG A169351-5MG A169351-250MG A169351-5G
77236-206EA 13 USD
77236-206 77236-208 77236-210 77236-212 77236-214 77236-216 77615-408 77615-410
Avibactam Sodium 98%
Avibactam Sodium

Precaución: for Research Use Only

Formula: C₇H₁₀N₃NaO₆S
MDL Number: MFCD28900719
CAS Number: 1192491-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR