2-Methoxy-6-methylbenzoic acid 98%

Supplier: Ambeed
Warning

A169319-100G A169319-10G A169319-1G A169319-250MG A169319-25G A169319-5G
77259-878EA 199.82 USD
77259-878 77259-880 77259-882 77259-884 77259-886 77259-888
2-Methoxy-6-methylbenzoic acid 98%
2-Methoxy-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.1700 g/mol
MDL Number: MFCD00463871
CAS Number: 6161-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR