3-(Methoxymethyl)benzoic acid 95%
Supplier: Ambeed
|
Warning
|
A169313-100MG
A169313-1G
A169313-250MG
77273-646EA
44.29
USD
77273-646
77273-648
77273-650
3-(Methoxymethyl)benzoic acid 95%
3-(Methoxymethyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₉H₁₀O₃ |
MDL Number:
MFCD01871355 CAS Number: 32194-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...